Orders LinkContactUsLinkComponent
BioCulture™ Pump, Electron Microscopy Sciences
BioCulture™ Pump, Electron Microscopy Sciences
BioCulture Calibration Head
Catalog # 102681-124
Supplier #:  65224-50
BioCulture™ Pump, Electron Microscopy Sciences
Catalog # 102681-124
BioCulture™ Pump, Electron Microscopy Sciences
BioCulture™ Pump, Electron Microscopy Sciences
BioCulture Calibration Head
Price
$1,971.34
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The BioCulture Calibration Head is A NIST traceable venture calibrator with 1% accuracy that allow users to calibrate the flow of the BioCulture pump themselves without having to send the pump out for service of calibration.
  • Allows flow calibration at flow rates between 30 – 120 LPM
  • Designed for use with impeller fan type pumps like the BioCulture™, which require calibration devices with very little backpressure resistance
Certifications: complies with a certificate of calibration traceable to NIST standards.
Specifications
  • Description:
    BioCulture Calibration Head