Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
INFINITY 4-11 Cameras, Electron Microscopy Sciences

INFINITY 4-11 Cameras, Electron Microscopy Sciences

INFINITY 4-11 Cameras, Electron Microscopy Sciences
Certificates
INFINITY 4-11 Cameras, Electron Microscopy Sciences
INFINITY 4-11 Cameras, Electron Microscopy Sciences

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

The INFINITY 4-11 camera series offers large format megapixel Kodak sensors for a wide field of view. The perfect choice for demanding high resolution imaging requiring excellent color rendition.

  • 11 megapixel resolution
  • Large format sensor
  • Superior light sensitivity with high fidelity color reproduction
  • Low noise electronics
  • F-mount lens adapter

Features include a 12-bit digital output, binning, progressive scan electronic shutter, full exposure control, auto white balance, programmable gain, sub-windowing and region of interest - ideal for high end ophthalmic, medical, clinical and life science applications.

Applications: Brightfield, Darkfield, DIC, Live Cell Imaging, Histology, Pathology, Cytology, Defect Analysis, Semiconductor Inspection, Metrology, Gel Documentation, and Moderate Light Fluorescence.

Specifications for Product Family

Specifications

  • Catalog No:
  • 102091-464
  • 102091-466
  • Model:
  • INFINITY4-11C
  • INFINITY4-11M
  • Sensor:
  • Color CCD
  • Monochrome CCD
  • Sensor size:
  • 35 mm
  • 35 mm
  • Pixels (W×H):
  • 4008×2672
  • 4008×2672
  • Frame Rate:
  • 3.5 fps
  • 3.5 fps
  • Interfaces:
  • USB 2.0
  • USB 2.0

Specifications

Product Family Options

Product Information

  • Resolution
    Sensor type
    Availability
    Price
  • 102091-464
    INFINITY4-11C Color Camera
    INFINITY4-11C Color Camera
    10.7 MP
    CCD
    Log in for availability
    Log in for availability
    Each
    Each - $19,848.67
    $19,848.67
    Each
       $19,848.67

    Supplier Number: 95114
    Specifications:
    More
    Specifications
    Cat. No.
    102091-464
    Model
    INFINITY4-11C
    Sensor
    Color CCD
    Sensor size
    35 mm
    Pixels (W×H)
    4008×2672
    Frame Rate
    3.5 fps
    Interfaces
    USB 2.0
  • 102091-466
    INFINITY4-11M Monochrome Camera
    INFINITY4-11M Monochrome Camera
    10.7 MP
    CCD
    Log in for availability
    Log in for availability
    Each
    Each - $19,848.67
    $19,848.67
    Each
       $19,848.67

    Supplier Number: 95115
    Specifications:
    More
    Specifications
    Cat. No.
    102091-466
    Model
    INFINITY4-11M
    Sensor
    Monochrome CCD
    Sensor size
    35 mm
    Pixels (W×H)
    4008×2672
    Frame Rate
    3.5 fps
    Interfaces
    USB 2.0

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.