Orders LinkContactUsLinkComponent
Micro Friedman Bone Rongeur, Electron Microscopy Sciences
Micro Friedman Bone Rongeur, Electron Microscopy Sciences
Micro Friedman Bone Rongeur
Catalog # 100494-552
Supplier #:  77981-01
Micro Friedman Bone Rongeur, Electron Microscopy Sciences
Catalog # 100494-552
Micro Friedman Bone Rongeur, Electron Microscopy Sciences
Micro Friedman Bone Rongeur, Electron Microscopy Sciences
Micro Friedman Bone Rongeur
Price
$464.84
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Bone Rongeur, curved. Very delicate. The jaw width is 1.3 mm. and it is 5 ½" long. Made from stainless steel with a polished finish.
Specifications
  • Description:
    Micro Friedman Bone Rongeur