ContactUsLinkComponent
Omniprobe Xtreme Access Short-Cut™ Probe Tips, Electron Microscopy Sciences
  102681-284
undefined
Omniprobe Xtreme Access Short-Cut™ Probe Tips, Electron Microscopy Sciences
  102681-284
 :  75960-06
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    XA Short Cut Tungsten Probe Tip
  • Cat. No.:
    102681-284
  • Supplier no.:
    75960-06
Custom tip designed from tungsten. Tip radius is < 0.5 µm with 6° taper angle for maximum lifetime service. Compatible with Short-Cut™ system for direct conversion to TEM grid
 : Xtreme Access Compatible Probe Point Holder is for use with 1/2" tungsten probe tips. Comes with a storage vessel. The AP250 AutoProbe™ tip holder uses the tungsten probe tip. Compatible with the Short-Cut™ for direct conversion to TEM Grid. Comes with a storage vial and handle.