Orders LinkContactUsLinkComponent
Tungsten Probe Tip, Electron Microscopy Sciences
Tungsten Probe Tip, Electron Microscopy Sciences
All Tungsten Probe Tip
Catalog # 102681-276
Supplier #:  75960-02
Tungsten Probe Tip, Electron Microscopy Sciences
Catalog # 102681-276
Tungsten Probe Tip, Electron Microscopy Sciences
Tungsten Probe Tip, Electron Microscopy Sciences
All Tungsten Probe Tip
Price
$392.37
Log in for availability
Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Custom dual taper tip designed from tungsten. Tip radius is 0.5 µm with 8 to 10° taper angle for maximum lifetime service.
  • For use with AutoProbe™ 200
  • Shank diameter is 0.508 mm (0.020")
  • Needle length is 34.036 mm (1.34")
The reduced taper angle allows for extended needle life for those users who cut and reshape their needles in the FIB.
Specifications
  • Description:
    All Tungsten Probe Tip