Orders LinkContactUsLinkComponent
Omniprobe TEM Grid and Sample Holders, Electron Microscopy Sciences
Omniprobe TEM Grid and Sample Holders, Electron Microscopy Sciences
TEM grid and sample holder, stainless steel
Catalog # 102681-310
Supplier #:  75969-SS
Omniprobe TEM Grid and Sample Holders, Electron Microscopy Sciences
Catalog # 102681-310
Omniprobe TEM Grid and Sample Holders, Electron Microscopy Sciences
Omniprobe TEM Grid and Sample Holders, Electron Microscopy Sciences
TEM grid and sample holder, stainless steel
Price
$2,518.32
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
TEM grid holder with stations for two TEM grids and two sample stubs. Available with two versions: very slightly magnetic stainless steel and non-magnetic aluminum. Long post – standard is 3.2 mm (1/8") diameter and 8.1 mm (0.32") length.
Delivery information: Comes with 2 sample stubs.
Specifications
  • Description:
    TEM grid and sample holder, stainless steel