Orders LinkContactUsLinkComponent
MAC Electron Backscatter Particle Analysis Standard, Electron Microscopy Sciences
Electron Backscatter Particle Analysis Standard
Catalog # 102099-008
 :  80070-EBP
MAC Electron Backscatter Particle Analysis Standard, Electron Microscopy Sciences
Catalog # 102099-008
CAS Number:  
MAC Electron Backscatter Particle Analysis Standard, Electron Microscopy Sciences
MAC Electron Backscatter Particle Analysis Standard, Electron Microscopy Sciences
Electron Backscatter Particle Analysis Standard
CAS Number:  
Price
$2,007.36
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Certified standards for system calibration in 13 mm aluminium pin stub.
  • Diamond polished to 0.25 um surface finish
  • Carbon Coated
This standard allows user to go across all grey areas.
Specifications
Specifications
  • Description:
    Electron Backscatter Particle Analysis Standard