Orders LinkContactUsLinkComponent
Large Set Screw Vises, Electron Microscopy Sciences
Large Set Screw Vises, Electron Microscopy Sciences
Large Set Screw Vises, Electron Microscopy Sciences
Certificates
Large Set Screw Vises, Electron Microscopy Sciences
Large Set Screw Vises, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This set screw vise is made from machined aluminum with stainless steel allen set screws has an open slot of diamension 10 W×5 D mm (0.394×0.197").
Product Family Options
Product Information
  • Availability
    Price
  • 102098-262
    Large set screw vise, 3.2 mm pin; dia. 25× H 10 mm
    Large set screw vise, 3.2 mm pin; dia. 25× H 10 mm
    Log in for availability
    Log in for availability
    Each
    Each - $62.49
    $62.49
    Each
       $62.49
    Supplier Number: 75941-03
  • 102098-264
    Large set screw vise, M4; dia. 25× H 10 mm
    Large set screw vise, M4; dia. 25× H 10 mm
    Log in for availability
    Log in for availability
    Each
    Each - $77.51
    $77.51
    Each
       $77.51
    Supplier Number: 75941-03H