Specifications
- Description:45° Pin Stub Holder, 3.2 mm (1/8") pin
- Cat. No.:102681-266
- Supplier no.:75952-45
Specifications
About this item
This 45 degree pin stub holder allows for a higher SE signal without having to tilt the specimen stage and it accommodates any standard 12.7 mm (1/2") pin stubs with 3.2 mm (1/8") pin. Grooved, machined aluminum with stainless steel Allen set screw.