Multi Pin Holder for 12 Pin Stubs, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
This 62 mm in diameter multi pin holder is designed to save time. It is made from machined aluminum with stainless steel Allen wrench screws.
It accommodates up to twelve standard 12.7 mm pin stubs.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.