Order Entry
Puerto Rico
ContactUsLinkComponent
Mounting Wax 52, Electron Microscopy Science
Mounting Wax 52, Electron Microscopy Science
Catalog # 101411-920
Minority or Woman-Owned Business Enterprise
CAS Number:  
undefined
Mounting Wax 52, Electron Microscopy Science
Catalog # 101411-920
Supplier Number:  50390-52
Minority or Woman-Owned Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • CAS No.:
    68955-36-2
  • Description:
    Mounting Wax 52
  • Size:
    100 g
  • Cat. No.:
    101411-920
  • Supplier no.:
    50390-52

Specifications

About this item

MWS052 is a tan colored wax, which is translucent on thin films. Softness makes it most useful with abrasive slurry cutting of materials. Best suited to non-porous surfaces such as glass or polished metal with a reasonably large area. Easily soluble in a variety of solvents.

113 Grams/1 Stick, 1" x 1" x 7.5" long (25 x 25 x 190 mm).
Soft Hardness and a Melting Point of 52°C.
Dilutent: Perchloroethylene