To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- CAS No.:68955-36-2
- Description:Mounting Wax 70
- Size:100 g
- Cat. No.:101411-922
- Supplier no.:50390-70
Specifications
About this item
Mounting Wax 70 is a quartz type sticky, orange colored wax (translucent in thin films) used to hold crystals or other materials while they are being sliced, lapped or polished. Moderate hardness and strength make it useful for bonding small samples in diamond wheel sawing and wire sawing operations. Widely used for holding semiconductors, optical materials, ceramics and metals with medium to large contact areas. Chemical resistant. 100 Gram /Pkg of 29 sticks, .25" dia x 4" long (6mm dia x 100mm). Moderate Hardness and a Melting Point of 70°C. Dilutent: Warm soapy water