Order Entry
Puerto Rico
ContactUsLinkComponent
Mounting Wax 80, Electron Microscopy Science
Mounting Wax 80, Electron Microscopy Science
Catalog # 101411-924
Minority or Woman-Owned Business Enterprise
CAS Number:  
undefined
Mounting Wax 80, Electron Microscopy Science
Catalog # 101411-924
Supplier Number:  50390-80
Minority or Woman-Owned Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • CAS No.:
    68955-36-2
  • Description:
    Mounting Wax 80
  • Size:
    25 g
  • Density:
    1.13 g/cm³ (9.43 lbs/gal)
  • Cat. No.:
    101411-924
  • Supplier no.:
    50390-80

Specifications

About this item

MWH080 is an amber colored wax, which is translucent in thin films.

  • Hard hardness with a Melting Point of 80 °C
  • Dilutent: Ethanol and Acetone
  • Chemical resistant

High hardness makes it ideal for sawing, ultrasonic cutting and lapping& polishing operations. Best suited to vitreous materials, polished metal surfaces, semiconductors and piezoelectric materials with large contacted areas.

Ordering information: 100 Gram/Pkg of 4 sticks .5" x .5" x 7" long (12.5 x 12.5 x 175mm).