Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- CAS No.:68955-36-2
- Description:Mounting Wax 100
- Size:20 g
- Cat. No.:101411-918
- Supplier no.:50390-100
Specifications
About this item
MWM100 Black Wax is designed for use as a masking material in jet polishing applications. As it is chemically inert, it is resistant to etching solutions such as hydrofluoric acid, perchloric acid and acetic acid. While it is resistant to many acids, it can be easily removed with hydrocarbon or chlorinated solvents.
100 Gram/Pkg (5 - 20 gram sticks).
Moderate Hardness and a Melting Point of 100° C.
Dilutent: Hydrocarbon or Chlorinated solvents