Orders LinkContactUsLinkComponent
Stage Micrometer Model SM-5, Electron Microscopy Sciences
Stage Micrometer Model SM-5, Electron Microscopy Sciences
Certificates
Stage Micrometer Model SM-5, Electron Microscopy Sciences
Stage Micrometer Model SM-5, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Linear Reference Scales are available in Chrome-on-Glass (G) and Chrome-on-Opal glass (O) or Chrome-on Quartz (Q)
Positive image are mostly clear or white.
 : The Opal and Quartz are offered as a special only and must be requested.
 : All EMS micrometers come in a sturdy wooden box.
Product Family Options
Product Information
  • Availability
    Price
  • 102680-932
    Stage Micrometer SM-5, Chrome on glass
    Stage Micrometer SM-5, Chrome on glass
    Log in for availability
    Log in for availability
    Each
    Each - $957.57
    $957.57
    Each
       $957.57
    Supplier Number: 60210-5PG
  • 102680-934
    Stage Micrometer SM-5 Chrome on Glass with Calibration
    Stage Micrometer SM-5 Chrome on Glass with Calibration
    Log in for availability
    Log in for availability
    Each
    Each - $2,473.70
    $2,473.70
    Each
       $2,473.70
    Supplier Number: 60210-5PGC