NGW1 Graticles, Electron Microscopy Sciences
About this item
Consists of 15 lines of equal length connecting the verticals of a regular hexagonal point network.
Reference: E.R.Weibel Lab. Invest. Vol. 22 pp 131-152. 'Principal and Methods for the Morphometric Study of the Lung and other Organs'.
3 Options Available Below
Product Details & Documents
Consists of 15 lines of equal length connecting the verticals of a regular hexagonal point network.
Reference: E.R.Weibel Lab. Invest. Vol. 22 pp 131-152. 'Principal and Methods for the Morphometric Study of the Lung and other Organs'.
Product Family Options
Product Information
- SizeAvailabilityPrice
- 100499-218NGW1 GraticuleSupplier Number: 68022-16
- 100499-220NGW1 GraticuleSupplier Number: 68022-19
- 100499-222NGW1 GraticuleSupplier Number: 68022-21