Orders LinkContactUsLinkComponent
EMS 9002 Vacuum Processor, Electron Microscopy Sciences
EMS 9002 Vacuum Processor, Electron Microscopy Sciences
EMS 9002 Vacuum Processor
Catalog # 102099-930
Supplier #:  97050-A
EMS 9002 Vacuum Processor, Electron Microscopy Sciences
Catalog # 102099-930
EMS 9002 Vacuum Processor, Electron Microscopy Sciences
EMS 9002 Vacuum Processor
Price
$3,902.80
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from Pyrex Glass, which offers better temperature conductivity and stability than any other material and is chemically resistant and microwave transparent.
The EMS 9002 when used in conjunction with our EMS 9000 processor improves ultra structural preservation during microwave assisted chemical fixation and reduces fixation and infiltration times dramatically.
Accessories information: The vacuum processor kit includes vacuum processor bowl, processor tray, vacuum processor tubing kit, and 74 cassette processing rack for processing of tissue samples in the microwave processor.
Specifications
  • Description:
    EMS 9002 Vacuum Processor
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • 74 Cassette Processing Rack
    74 Cassette Processing Rack
    103306-386
    Log in for availability
    Each - $591.14
Product Information
  • Catalog #
    Availability
    Price
  • Vacuum Processing Bowl
    Vacuum Processing Bowl
    103306-388
    Log in for availability
    Each - $601.72
Product Information
  • Catalog #
    Availability
    Price
  • Non-Vacuum Processor Kit
    Non-Vacuum Processor Kit
    103306-330
    Log in for availability
    Each - $1,650.47
Product Information
  • Catalog #
    Availability
    Price
  • Processor Tray
    Processor Tray
    103305-782
    Log in for availability
    Each - $1,652.45
Product Information
  • Catalog #
    Availability
    Price
  • Vacuum Processor Kit
    Vacuum Processor Kit
    103306-936
    Log in for availability
    Pack of 1 - $1,815.24
Product Information
  • Catalog #
    Availability
    Price
  • Vacuum Processor Tubing Kit
    Vacuum Processor Tubing Kit
    103302-366
    Log in for availability
    Each - $59.54
Product Information
  • Catalog #
    Availability
    Price
  • Non-Vacuum Processing Bowl
    Non-Vacuum Processing Bowl
    103305-560
    Log in for availability
    Each - $390.18