Orders LinkContactUsLinkComponent
Sure-Footed Biodegradable Beaker, Electron Microscopy Sciences
Sure-Footed Biodegradable Beaker, Electron Microscopy Sciences
250 ml (8.5 oz.), Sure-Footed Beaker, Biodegradable, 250 ml
Catalog # 102091-118
Supplier #:  68553
Sure-Footed Biodegradable Beaker, Electron Microscopy Sciences
Catalog # 102091-118
Sure-Footed Biodegradable Beaker, Electron Microscopy Sciences
Sure-Footed Biodegradable Beaker, Electron Microscopy Sciences
250 ml (8.5 oz.), Sure-Footed Beaker, Biodegradable, 250 ml
Price
$140.12
Log in for availability
Pack of 50
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Innovative outer beaker base ensures stability with small volumes of liquid.
  • 100% Biodegradable - Made from the same environmentally conscious Bio-PET as our Bio-Pure reservoirs
  • Thermally Insulating - Air void allows beakers to hold liquid temperatures 30% longer than standard polypropylene beakers
  • Unique Pipet Rest - Diametrically opposed grooves act as a pipet rest when not in use
Alternate Uses:
The thermal retentive characteristics of the Sure-Footed Beaker allow it to be used as a small bucket for ice/dry ice.
Allow beaker to sink in a water bath by piercing or melting a ventilation hole in the beaker rim.
Beakers may be covered or piggybacked to increase thermal insulation.
Specifications
  • Capacity:
    250 ml (8.5 oz.)
  • Description:
    Sure-Footed Beaker, Biodegradable, 250 ml