Specifications
- Description:Frozen Section Brushes, set of 4
- Cat. No.:102093-794
- Supplier no.:61735-01
Specifications
About this item
The stiffness of this bristle will prevent the frozen section from curling through the brush. The flat white boar bristle brush is cut into an angle.
Set of four brushes: two #1 with ⅜" angle cut and #2 with ¼" angle cut.