Orders LinkContactUsLinkComponent
Karnovsky Fixative Reagent Grade, Electron Microscopy Sciences
Karnovsky Fixative Reagent Grade, Electron Microscopy Sciences
Karnovsky Fixative Reagent Grade, Electron Microscopy Sciences
Karnovsky Fixative Reagent Grade, Electron Microscopy Sciences
Fixative Reagent Grade (Karnovsky’s). This fixative contains 3% glutaraldehyde and 2% formaldehyde in 0.1M phosphate buffer, pH 7.4. Karnovsky’s with formalin 2% and Biological glutaraldehyde 3% in 0.1m Phosphate Buffer. Karnovsky’s with paraformaldehyde solution and EM grade glutaraldehyde.
Product Family Options
  • Description
    Size
  • Karnovsky Fixative Reagent Grade
    1 L
    Each
    Each - $562.73
    $562.73
    Each
       $562.73
     : 15731-10
  • Karnovsky Fixative Paraformaldehyde Solution and glut
    1 L
    Each
    Each - $617.25
    $617.25
    Each
       $617.25
     : 15732-10