ContactUsLinkComponent
Universal Calibration Slide PS20, Electron Microscopy Sciences
Universal Calibration Slide PS20, Electron Microscopy Sciences
Catalog # 102096-154
undefined
Universal Calibration Slide PS20, Electron Microscopy Sciences
Catalog # 102096-154
Supplier Number:  68053-20
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Species:
  • Description:
    Universal calibration slides PS20
  • Environmentally Preferable:
  • Cat. No.:
    102096-154
  • Supplier no.:
    68053-20
Specifications
About this item
Multiple images on a slide provide the most cost-effective solution to calibration and resolution checking of microscopes and image analysis systems
Calibration of microscopes and image analysis systems is becoming more sophisticated, with the requirement being for a variety of image patterns to satisfy the numerous parameters. We’re now introducing a new multi-function calibration standard specifically for these applications.
Multiple images on a slide provide the most cost-effective solution to calibration and resolution checking of microscopes and image analysis systems. The combination of scales, dots, circles, squares, rulings, grids and angles can be supplied with an internationally traceable certificate of calibration for those who require ISO conformity. Each glass slide has a permanent serial number and can be supplied with full or partial UKAS certificate of accuracy. Starting from a fixed ʽDatum Point’ mark, each individual pattern or array can be located using X, Y, coordinates.
The combination of scales, dots, circles, squares, rulings, grids and angles can be supplied with an internationally traceable certificate of calibration for those who require ISO conformity. Each glass slide has a permanent serial number and can be supplied with full or partial UKAS certificate of accuracy. Starting from a fixed ʽDatum Point’ mark, each individual pattern or array can be located using X, Y, coordinates.