About this item
PS25 Pattern Detail: Diamonds: (Point to point) 0.5, 0.3, 0.2, 0.1
Reflected light calibration slide with diamond shapes and scales for calibrating Vickers and Rockwell hardness testers. For Vickers and Rockwell methods we offer the PS25 which has a series of diamond shapes of varying size and x-y scales. Each of the markings on the slide is clearly identified with its size. The PS25 has a glass disc with the image precision marked in vacuum deposited chrome and this is cemented into a stainless steel slide mount, making the item very durable. The slide has a unique serial number indelibly marked on the slide mount and can be supplied with an Internationally traceable certificate of calibration. This product is supplied in a polished wooden box.
PS25 Pattern Detail: Diamonds: (Point to point) 0.5, 0.3, 0.2, 0.1. 0.05. and 0.02 mm
Scales: Horizontal & Vertical, 2 mm in 0.1 mm divisions
0.05. and 0.02 mm