ContactUsLinkComponent
Standard Pin SEM Mounts with Slotted Numbered Head, Electron Microscopy Sciences
Standard Pin SEM Mounts with Slotted Numbered Head, Electron Microscopy Sciences
Catalog # 102098-148
undefined
Standard Pin SEM Mounts with Slotted Numbered Head, Electron Microscopy Sciences
Catalog # 102098-148
Supplier Number:  75220-12-500
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Pin Stub Slotted Head Numbered
  • Material:
    Aluminum
  • Cat. No.:
    102098-148
  • Head diameter:
    12.7 mm
  • Supplier no.:
    75220-12-500
  • Pin size:
    8 H×3.2 Ø mm
Specifications
About this item
Slotted Numbered head pin stub mounts are for use with Cambridge, Leica, LEO, ZEISS, Philips, FEI, CamScan, Tescan, ETEC. Mounts made from lathe finished ultra-pure aluminum. The head is numbered 1-alpha/4-numeric.
Ordering information: Designate number sequence when ordering.