ContactUsLinkComponent
Set Screw Vise, Electron Microscopy Sciences
Set Screw Vise, Electron Microscopy Sciences
Catalog # 102098-260
undefined
Set Screw Vise, Electron Microscopy Sciences
Catalog # 102098-260
Supplier Number:  75941-01
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Set Screw Vise, 1/2" dia.
  • Cat. No.:
    102098-260
  • Supplier no.:
    75941-01
Specifications
About this item
This set screw vice which is made from machined aluminum with stainless steel allen set screws has an open slot that is 4 mm wide x 5 mm deep. And has a 3.2 mm dia. pin.