Orders LinkContactUsLinkComponent
Freezing Griddle/ Elevated Freezing Block Set, Electron Microscopy Sciences
Freezing Griddle/ Elevated Freezing Block Set, Electron Microscopy Sciences
Freezing Griddle Elevated Freezing Block Set
Catalog # 102093-748
Supplier #:  61706-01
Freezing Griddle/ Elevated Freezing Block Set, Electron Microscopy Sciences
Catalog # 102093-748
Freezing Griddle/ Elevated Freezing Block Set, Electron Microscopy Sciences
Freezing Griddle/ Elevated Freezing Block Set, Electron Microscopy Sciences
Freezing Griddle Elevated Freezing Block Set
Price
$855.32
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Block set includes cutting board/freezing griddle, elevated freezing block, epoxy coated forceps, putty knife, extra elevating bars, and cytology funnel sample chambers.
Cutting board/Freezing griddle: The steel “freezing griddle” serves as a freezing surface on which to prepare frozen blocks when used in conjunction with the elevated freezing block. The small attached cutting board serves as a cold surface to cut the frozen block while maintaining the freezing temperature. This simple piece of apparatus is kept cold in the cryostat and is removed to the work bench only during the process of cutting the frozen block. As sections are cut, they are kept cold on the freezing griddle.
Elevated Freezing Block: This steel block is kept cold in the cryostat and in used in conjunction with the freezing griddle. The two broad sides of the block are equipped with feet measuring 3.0 mm on one side and 4.5 mm on the other. The feet create a space between the two steel bars in which the frozen blocks are prepared. Additional elevating bars are supplied to further elevate the block when preparing very thick portion of tissue.
Specifications
  • Description:
    Freezing Griddle Elevated Freezing Block Set