ContactUsLinkComponent
Wooden Storage Box for Pin Mounts, Electron Microscopy Sciences
Wooden Storage Box for Pin Mounts, Electron Microscopy Sciences
Catalog # 102091-320
undefined
Wooden Storage Box for Pin Mounts, Electron Microscopy Sciences
Catalog # 102091-320
Supplier Number:  76507
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Wooden Storage Box for Pin Mounts
  • Cat. No.:
    102091-320
  • Supplier no.:
    76507
Specifications
About this item
EMS offers a high end wooden storage box for the storage of 12.7 mm (1/2") pin mounts.
The box can store up to 128 mounts in rubber foam with complete indexed sides.