Orders LinkContactUsLinkComponent
Lab-Sorb, Electron Microscopy Sciences
Lab-Sorb, Electron Microscopy Sciences
Lab-Sorb, Electron Microscopy Sciences
Lab-Sorb, Electron Microscopy Sciences
Converts Liquid Waste to Solid Waste for Easy Disposal
Convenient - Includes extra durable disposal bags, super absorbant LabSorb granules and a convenient graduated scoop.
Prevents Spills - Reduces the possibility of accidentally spilling liquid waste in the lab. If spills do occur, use LabSorb for safe liquid containment during cleanup.
Quick - Convert liquids to solids in seconds.
Adaptable - Apply to any volume of liquid waste including 500 mL, 600 mL, 1L and 2L vessels. One jar of LabSorb can absorb up to 50 litres of liquid.
Versatile - LabSorb granules can rapidly absorb aqueous or mixed aqueous and organic solvent waste
Product Family Options
  • Description
    Capacity
  • Lab-Sorb 16 oz jar
    16 oz.
    Each
    Each - $73.30
    $73.30
    Each
       $73.30
     : 68552-06
  • Kit Lab-Sorb 16 oz jar plus 25 small bags
    Each
    Each - $88.08
    $88.08
    Each
       $88.08
     : 68552-07
  • Kit Lab-Sorb 16 oz jar plus 25 medium bags
    Each
    Each - $88.08
    $88.08
    Each
       $88.08
     : 68552-08
  • Kit Lab-Sorb 16 oz jar plus 25 large bags
    Each
    Each - $92.02
    $92.02
    Each
       $92.02
     : 68552-09
  • Variety kit Lab-sorb plus variety size bags
    Each
    Each - $92.02
    $92.02
    Each
       $92.02
     : 68552-10
  • 50 small bags for Lab-sorb
    Pack of 50
    Pack of 50 - $55.05
    $55.05
    Pack of 50
       $55.05
     : 68552-11
  • 50 medium bags for Lab-sorb
    Pack of 50
    Pack of 50 - $52.86
    $52.86
    Pack of 50
       $52.86
     : 68552-12
  • 50 large bags for Lab-sorb
    Pack of 50
    Pack of 50 - $52.86
    $52.86
    Pack of 50
       $52.86
     : 68552-13