About this item
This light and compact passive platform has been specially designed for instruments such as microscopes.
- Made entirely from electroless nickel plated aluminium parts it is suitable for class 1 requirements
- The top plate is 1 cm thick with three damped isolators mounted to the underside
Its T-shaped profile allows users to place their arms on either side of the platform while the width at the rear is large enough to support a wide range of instruments.