About this item
ProCrystal™ Protein Crystallization Covers: Hanging drop microtiter plate seals with unmatched drop formation and segregation and no noticeable x-ray diffraction.
The seals sit directly over the microplate crystallization wells. The seals are validated to be incorruptible and materials will not ingress into samples. They are available in either an individual well pre-cut format so that one can easily remove crystals using a forceps from individual wells without affecting neighboring wells or uncut format. The plate seals are UV compatible with the highest optical quality available. They are exceptionally hydrophobic which maintains drop footprint and segregation even with the use of MPD, Glycerol or detergents.