ProPlate™ Microtiter Plate, Electron Microscopy Sciences
About this item
Patented ProPlate™ technology for high throughput robotic processing of microarrays
The proprietary adhesive forms a water-tight seal between a microarray substrate and the bottomless microtiter plate. Grace Bio Labs offers plate with SBS standard spacing in both 96-well (shown) and 384-well formats.
Key product features include:
Guaranteed flat SBS compliant microtiter plate format which enables robotic processing of assays
Water-tight seal between superstructure and substrate
Compatible with glass or plastic substrates
Available in 96 round or square or 384 square well format
Available as a complete microtiter plate with microcellulose film coated substrate, uncoated substrate or as an adhesive superstructure alone
Plates are available with SBS standard spacing in both 96-well and 384-well formats
Grace BioLabs developed bottomless SBS compliant microtiter plate superstructures for use with robotic processors. TheSuperstructures come in a permanently affixed well format – the ProPlate MP™, or a removable well format – the FlexPlate MP™. The ProPlate MP™ SBS compliant microtiter plate features a SecureSeal™ Plus adhesive bonding system applied to a bottomless 96-round well, black, polystyrene microtiter plate. Simply peel off the protective polyliner and apply large format array for automated processing.
13 Options Available Below
Product Details & Documents
Patented ProPlate™ technology for high throughput robotic processing of microarrays
The proprietary adhesive forms a water-tight seal between a microarray substrate and the bottomless microtiter plate. Grace Bio Labs offers plate with SBS standard spacing in both 96-well (shown) and 384-well formats.
Key product features include:
Guaranteed flat SBS compliant microtiter plate format which enables robotic processing of assays
Water-tight seal between superstructure and substrate
Compatible with glass or plastic substrates
Available in 96 round or square or 384 square well format
Available as a complete microtiter plate with microcellulose film coated substrate, uncoated substrate or as an adhesive superstructure alone
Plates are available with SBS standard spacing in both 96-well and 384-well formats
Grace BioLabs developed bottomless SBS compliant microtiter plate superstructures for use with robotic processors. TheSuperstructures come in a permanently affixed well format – the ProPlate MP™, or a removable well format – the FlexPlate MP™. The ProPlate MP™ SBS compliant microtiter plate features a SecureSeal™ Plus adhesive bonding system applied to a bottomless 96-round well, black, polystyrene microtiter plate. Simply peel off the protective polyliner and apply large format array for automated processing.
Product Family Options
Product Information
- AvailabilityPrice
- 102094-72896 Square Well Proplate Adhesive SBS Microtiter Plate, Black Polystyrene96 Square Well Proplate Adhesive SBS Microtiter Plate, Black PolystyreneLog in for availabilityEach$104.73Supplier Number: 63485-01
- 102094-73096 Round Well Proplate Adhesive SBS Microtiter Plate, Black Polystyrene96 Round Well Proplate Adhesive SBS Microtiter Plate, Black PolystyreneLog in for availabilityEach$125.08Supplier Number: 63485-02
- 102094-732364 Square Well Proplate Adhesive SBS Microtiter Plate, Black Polystyrene364 Square Well Proplate Adhesive SBS Microtiter Plate, Black PolystyreneLog in for availabilityEach$143.05Supplier Number: 63485-05
- 102094-73496 Square FlexPlate, Adhesive 1 Side, Clear Silicone, 7.25x7.25 mm96 Square FlexPlate, Adhesive 1 Side, Clear Silicone, 7.25x7.25 mmLog in for availabilityEach$105.54Supplier Number: 63485-10
- 102094-73696 Square FlexPlate, Adhesive 1 Side, 7.25x7.25 mm, Black Polystyrene Frame96 Square FlexPlate, Adhesive 1 Side, 7.25x7.25 mm, Black Polystyrene FrameLog in for availabilityEach$168.21Supplier Number: 63485-11
- 102094-738ONCYTE AVID® 96 NC Round Pad, Glass SubstrateSupplier Number: 63485-20
- 102094-740ONCYTE AVID® 96 NC Square Pad, 6x6 mm, Glass SubstrateSupplier Number: 63485-21
- 102094-742ONCYTE AVID® 96 NC Round Pad, 6 mm dia, Plastic SubstrateSupplier Number: 63485-22
- 102094-744ONCYTE AVID® 384 NC Square Pad, 2.5x2.5 mm, Glass SubstrateSupplier Number: 63485-23
- 102094-746ONCYTE AVID® 96 NC Round Pad, 6 mm dia, Glass SubstrateSupplier Number: 63485-24
- 102094-748ONCYTE AVID® 96 NC Square Pad, 6x6 mm, Glass SubstrateSupplier Number: 63485-25
- 102094-750ONCYTE AVID® 96 NC Round Pad, 6 mm dia, Plastic SubstrateSupplier Number: 63485-26
- 102094-752ONCYTE AVID® 384 NC Square Pad, 2.5x2.5 mm, Glass SubstrateSupplier Number: 63485-27