Order Entry
Puerto Rico
ContactUsLinkComponent
FlowMesh Sheet, Electron Microscopy Sciences
FlowMesh Sheet, Electron Microscopy Sciences
Catalog # 102681-178
undefined
FlowMesh Sheet, Electron Microscopy Sciences
Catalog # 102681-178
Supplier Number:  68505-20

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    FlowMesh Sheet 8.50 x 11.00" / 216 mm x 279 mm
  • Cat. No.:
    102681-178
  • Supplier no.:
    68505-20

Specifications

About this item

A Permeable Support Mesh for Gels and Membranes

  • Protects - Place FlowMesh above or below a gel or filter to protect from tearing, bending or folding
  • Sturdy - Supports gels during incubation with nucleic acid probes, antibody probes, equilibration buffers, stain, destain solutions, etc.
  • Versatile - Use as a permeable spacer for stacking as well as for staining and destaining procedures
  • Access - Both sides of a container or plastic bag are assured of constant solution access, which improves the uniformity of hybridizations and binding assays.
  • Convenient - FlowMesh is re-usable and easily cut to size