ContactUsLinkComponent
Marking Labels "Bake Oven-Safe", Electron Microscopy Sciences
Marking Labels "Bake Oven-Safe", Electron Microscopy Sciences
  101413-850
Minority or Woman-Owned Business Enterprise
undefined
Marking Labels "Bake Oven-Safe", Electron Microscopy Sciences
  101413-850
 :  77540-W
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Color:
    White
  • Description:
    Bake Tough-Tags
  • Environmentally Preferable:
  • Cat. No.:
    101413-850
  • Supplier no.:
    77540-W
Ideal use with oven sterilization. Resists oven temperatures up to 180 °C, autoclavable, boilable and solvent resistant and heating to inactivate RNase, DNase and other enzymes.
  • Adhere to any surface
  • Bake Tags withstand cryogenic freezer storage conditions (liquid nitrogen)
  • Withstand boiling water baths without experiencing any problems
Unlike conventional labels that become brittle, darken and fall off bottles, beakers, pipet canisters and other containers in the oven, bake tags resist most organic solvents and caustic agents, e.g. alcohols, most aromatic and some chlorinated solvents as well as acids and alkalis.