Specifications
- Description:Autoradiography ruler
- Environmentally Preferable:
- Cat. No.:102096-324
- Supplier no.:68550-04
Specifications
About this item
The Autoradiography Ruler Set (ARS) is an orientation and measurement device that exposes metric ruler markings directly onto X-ray film.
- Phosphorescent, non-radioactive metric rulers for radiographs
- Make easy electrophoretic mobility calculations
Utilizes a specially formulated phosphorescent dye compound and a unique emission filter which serves as an ideal non-radioactive substitute for ink markings. The phosphorescent emission of each ruler can be regenerated indefinitely and can be reused any number of times. Produces a high resolution image that is unaffected by intensifying screens or pre-flashed film.