Orders LinkContactUsLinkComponent
Microtube Rack - Microwaveable, Electron Microscopy Sciences
Microtube Rack - Microwaveable, Electron Microscopy Sciences
Microtube Rack - Microwaveable, Electron Microscopy Sciences
Certificates
Microtube Rack - Microwaveable, Electron Microscopy Sciences
Microtube Rack - Microwaveable, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Polypropylene floating racks keep microtubes submerged in a water bath – perfect for controlling sample temperature. Detachable legs give the users more options: attach legs so rack stands upright on benchtop, or detach legs so sample tubes can be quickly popped out by pushing down onto any surface. Ideally to hold microcentrifuge tube in water bath during polymerization, such as LR White®. Not autoclavable. Measures: 60 mm high including legs.
Product Family Options
Product Information
  • Availability
    Price
  • 101454-124
    Rack Micro Tube Sq white
    Rack Micro Tube Sq white
    Log in for availability
    Log in for availability
    Each
    Each - $60.05
    $60.05
    Each
       $60.05
    Supplier Number: 72372-SW
  • 101454-118
    Rack Micro Tube Rnd white
    Rack Micro Tube Rnd white
    Log in for availability
    Log in for availability
    Each
    Each - $55.96
    $55.96
    Each
       $55.96
    Supplier Number: 72372-R20
  • 101454-122
    Rack Micro Tube Sq black
    Rack Micro Tube Sq black
    Log in for availability
    Log in for availability
    Each
    Each - $53.02
    $53.02
    Each
       $53.02
    Supplier Number: 72372-SB
  • Discontinued
    Supplier Number: -