Orders LinkContactUsLinkComponent
Calibration Micrometer, Electron Microscopy Sciences
Calibration Micrometer, Electron Microscopy Sciences
Calibration Micrometer, Electron Microscopy Sciences
Certificates
Calibration Micrometer, Electron Microscopy Sciences
Calibration Micrometer, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This ideal standard provides accurate calibration of any eyepiece, reticules, filar micrometers, or electronic measuring instrument
Features a series of ʽH’ shaped fiducial images in a range of sizes from 0.1 mm through 20 mm, useful for calibration range from 1X to 1000X power.
Product Family Options
Product Information
  • Availability
    Price
  • 102093-154
    Model ACM-1, Calibrating Micrometer
    Model ACM-1, Calibrating Micrometer
    Log in for availability
    Log in for availability
    Each
    Each - $661.13
    $661.13
    Each
       $661.13
    Supplier Number: 60218-1PG
  • 102680-948
    Model ACM-1, with 11 pts Calibrated
    Model ACM-1, with 11 pts Calibrated
    Log in for availability
    Log in for availability
    Each
    Each - $1,517.46
    $1,517.46
    Each
       $1,517.46
    Supplier Number: 60218-1PGC