ContactUsLinkComponent
Vibrating Microtome Tissue Slicer Options, Electron Microscopy Sciences
Vibrating Microtome Tissue Slicer Options, Electron Microscopy Sciences
Catalog # 102100-464
Vibrating Microtome Tissue Slicer Options, Electron Microscopy Sciences
Catalog # 102100-464
Supplier Number:  EMS7000SMZ-2
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Programable Vibrating Microtome
  • Cat. No.:
    102100-464
  • Electrical:
    115 V, 60 Hz or 230 V, 50 Hz
  • Supplier no.:
    EMS7000SMZ-2
Specifications
About this item
The 7000smz-2 unit with a Z-axis deflection of Sub-Micron and a blade advance controllable to 10 microns/second
The all-new user interface is both easy to use and versatile. The 7000smz-2 and both 5100mz models offer similar operation at the press of a button. The versatility includes simple operation at the push of a button or a range of changeable and programmable parameters and a menu enables the settings of your own preferences. The 7000smz-2 also gives you longevity of performance; in other words it not only gives submicron performance out-of-the-box, but the advance vibrating mechanism does not contain bearings and other components subject to wear. Consequently it will retain the sub-micron Z-axis deflection performance for years to come, giving you consistency in your biological preparation. Tissue cooling is accomplished by the use of either an ice-water bath or an electronically comtrolled thermo-electric cooler. Other options include a LED cold light source and a magnifier or a stereoscope for clear observation whilst slicing. The new 5100mz series Vibrating Microtomes share the user interface with the premium 7000smz-2, placing a wide range of adjustable parameters at the user's fingertips. Use in fully manual or the semi-automated "slice window" mode which automatically remembers the start and finish position of the slice.
The 5100-Plus is perfect for those who need to keep slices viable for longer, e.g. for electrophysiological field recordings. The user can calibrate the Z-axis deflection of the blade to 2µm with the adjustable blade holder and "optical" calibration device. Minimal Z-axis deflection reduces the "chatter" and minimizes damage to the slice surface, leading to greater viability of the cells on the slice surface.The 5100mz is a very competitively priced, high precision, vibrating microtome which shares many features with the top of the range 7000smz series, such as the vibrating mechanism, the inner and outer tissue baths and the easy to use control system. This new entry level microtome is perfect for techniques such as histology, organotype slice culture and low resolution imaging with a fixed blade holder and a mechanism Z-axis error rate of 5-8µm.
The 5100mz-Plus and 5100mz are very competitively priced high precision units with a Z-axis deflection of only 1-2 microns and a blade advance controllable to 10 microns/sec. On all units the section thickness step size is 0.001mm. The 7000smz-2 and 5100mz-Plus are supplied with their own Z-axis calibration verifier. All types of sectioning is possible including sectioning for visual patching of neurological tissue, heart, and lung, and much more. The effect of excessive Z-axis deflection on the health and viability of the tissue preparation has been much discussed since the publication of Jonas et al (2003) and the 7000smz-2 will now deliver perfect sections every time with sub-micron Z-axis deflection across a wide range of vibration speeds and amplitudes. The affordable 5100mz-Plus offers almost the same features with Z-axis deflection of 1-2 microns at all vibration speeds and amplitudes, which is less than most other microtomes on the market.
Ordering information: The 7000smz-2 features a Z-axis calibration unit, minimal Z-axis deflection (less than 1µm) at all speeds and amplitudes, Z-axis blade adjust minimizer, blade holder angle to user requirement, set start and stop position of blade travel.