Orders LinkContactUsLinkComponent
mini-BUCK™ Primary Flow Calibrator, Electron Microscopy Sciences
mini-BUCK™ Primary Flow Calibrator, Electron Microscopy Sciences
mini-BUCK™ Primary Flow Calibrator, Electron Microscopy Sciences
Certificates
mini-BUCK™ Primary Flow Calibrator, Electron Microscopy Sciences
mini-BUCK™ Primary Flow Calibrator, Electron Microscopy Sciences
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The mini-BUCK™ Primary Flow Calibrator is the industry standard for day-to-day flow verification and calibration of your personal and environmental sampling pumps.
  • No more lugging oversized calibration panels
  • No more rotameter inaccuracies
It is also suitable for verifying rotameters, setting instrument panels, and calibrating laboratory instruments. The mini-BUCK™ is accurate at any attitude and can be used for flow measurements with sampling media.
Product Family Options
Product Information
  • Flow rate
    Availability
    Price
  • 102681-152
    Low Flow Calibrator, 120 V
    Low Flow Calibrator, 120 V
    0.1 - 300 cc/min
    Log in for availability
    Log in for availability
    Each
    Each - $2,618.15
    $2,618.15
    Each
       $2,618.15
    Supplier Number: 65240-60
  • 102681-154
    Medium Flow Calibrator, 120 V
    Medium Flow Calibrator, 120 V
    1- 6000 cc/min
    Log in for availability
    Log in for availability
    Each
    Each - $2,252.04
    $2,252.04
    Each
       $2,252.04
    Supplier Number: 65240-61
  • 102681-156
    High Flow Calibrator, 120 V
    High Flow Calibrator, 120 V
    100 cc/min - 30 LPM
    Log in for availability
    Log in for availability
    Each
    Each - $2,252.04
    $2,252.04
    Each
       $2,252.04
    Supplier Number: 65240-63
  • 102681-158
    High Flow Calibrator with 1/2" port, 120 V
    High Flow Calibrator with 1/2" port, 120 V
    100 cc/min - 30 LPM
    Log in for availability
    Log in for availability
    Each
    Each - $2,109.45
    $2,109.45
    Each
       $2,109.45
    Supplier Number: 65240-64