Specifications
- Description:Sensidyne Gilian BDXII Pump
- Cat. No.:102681-160
- Supplier no.:65241-12
Specifications
About this item
The Gilian BDX II Abatement Air Sampler from Sensidyne is a dependable, low cost personal sampling pump.
- Flow rates – 500 – 3000 cc/min ( 0.50-3 LPM)
- Rechargeable battery pack system
- Built-in rotameter & flow adjust
- Compact, lightweight, quiet
- Full one year warranty
It is a “no-frills” model designed for use with asbestos and lead sampling. The Gilian BDX II includes a Pump with battery pack, battery charger, tool kit, 36” tubing, clip assembly and manual.