Orders LinkContactUsLinkComponent
Cassette Opener, Electron Microscopy Sciences
Cassette Opener, Electron Microscopy Sciences
Cassette Opener
Catalog # 102681-132
Supplier #:  65231-01
Cassette Opener, Electron Microscopy Sciences
Catalog # 102681-132
Cassette Opener, Electron Microscopy Sciences
Cassette Opener, Electron Microscopy Sciences
Cassette Opener
Price
$105.13
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Heavy duty stainless steel cassette opener. Works with 25 mm, 37 mm, and Air-O-Cell cassettes. No need to remove labels, this opener cuts right through and effortlessly pries cassettes open.
Specifications
  • Description:
    Cassette Opener