ContactUsLinkComponent
Zefon Bio-Pump® Plus, Electron Microscopy Sciences
Zefon Bio-Pump® Plus, Electron Microscopy Sciences
Catalog # 102681-060
undefined
Zefon Bio-Pump® Plus, Electron Microscopy Sciences
Catalog # 102681-060
Supplier Number:  65211-12
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Flow rate:
    15 LPM
  • Description:
    Zefon Bio-Pump Plus
  • Cat. No.:
    102681-060
  • Supplier no.:
    65211-12
Specifications
About this item
The Zefon Bio-Pump Plus® is the smallest and lightest portable battery powered IAD sampling pump available.
The Zefon Bio-Pump Plus® has Nickel metal hydride battery which withstand upto 100 samples. A quick-charge battery charger (approx. 3 hours) also included.
It provides the simplest and most convenient way to sample with Air-O-Cell® and Via-Cell® cassettes at a flow rate of 15 LBM. Each pump come complete with both an Air-O-Cell® and Via-Cell® flow indicator, quick charger, 10 free Air-O-Cell ® cassettes and a handy carrying case.
Weight: 1.6 lbs.