ContactUsLinkComponent
Epo-Tek® H22 Adhesive, Electron Microscopy Sciences
  102091-814
 :  
undefined
Epo-Tek® H22 Adhesive, Electron Microscopy Sciences
  102091-814
 :  12673-22
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    H22  Adhesive
  • Size:
    1 oz
  • Cat. No.:
    102091-814
  • Supplier no.:
    12673-22
Epo-Tek® H22 is a two component, silver-filled epoxy system.
  • Smooth, free-flowing, slightly thixotropic paste
  • Excellent resistance to solvents, chemicals and moisture
  • Extended pot life and fast curing at low temperature 100°C
  • Designed for die bonding and sealing hybrid circuit
Adhesive's high Tg allows it to be used for high temperature applications. It contains no solvents; it is a NASA approved low outgasing epoxy. Recommended for SEM small angle cleavage and wafer bonding. Mixing ratio of silver resin paste and liquid hardener is 100:4.5. Pot life is 16 hours and shelf life is 6 months at room temperature.