TEM sample grid coupons designed specially for use in the Short-Cut™ tool. Available in two versions: One made from pure copper and the other is molybdenum coated copper.
- Frontside Thinning, 45°, 45° cut
- Backside Thinning, 45°, 45° cut
- Frontside Thinning, 26.5°, 26.5° cut
- Backside Thinning, 26.5°, 26.5° cut
Frontside Thinning, 26.5°, the center portion is a standard 3 mm grid into which the sample loaded needle is swaged and cut 26.5°.
Backside Thinning, 26.5°, the center portion is a standard 3 mm grid into which the sample loaded needle is swaged and cut 26.5°.
Frontside Thinning, 45°, the center portion is a standard 3 mm grid into which the sample loaded needle is swaged and cut 45°.
Backside Thinning, 45°, the center portion is a standard 3 mm grid into which the sample loaded needle is swaged and cut 45°.