Omniprobe Omni Grid Storage Box, Electron Microscopy Sciences
102681-302
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Description:Grid Storage Box
- Cat. No.:102681-302
- Supplier no.:75965-01
Storage box for 100 standard or haft grids.
- Measures 3 mm in diameter
- TEM grids Box comes complete with base, lid and clips