Omniprobe Lift-Out Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
Custom copper lift-out grids specifically designed for in-situ lift-out.
- Multiple indexed mounting locations
- With vertical bars and “V” shaped attachment surfaces
Measures 3 mm in diameter.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.