Orders LinkContactUsLinkComponent
Tungsten/Nickel Probe Tips, Electron Microscopy Sciences
Tungsten/Nickel Probe Tips, Electron Microscopy Sciences
Tungsten/Nickel Probe Tip
Catalog # 102681-274
Supplier #:  75960-01
Tungsten/Nickel Probe Tips, Electron Microscopy Sciences
Catalog # 102681-274
Tungsten/Nickel Probe Tips, Electron Microscopy Sciences
Tungsten/Nickel Probe Tips, Electron Microscopy Sciences
Tungsten/Nickel Probe Tip
Price
$474.21
Log in for availability
Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Custom probe tip with a nickel tube shank and tungsten tip for use with Omniprobe systems. Tip radius is 0.5 µm with 13° taper angle for maximum lifetime service.
Specifications
  • Description:
    Tungsten/Nickel Probe Tip