Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Decon 90 Cleaning Agent, Electron Microscopy Sciences

Decon 90, 1 l, Bottle
  102093-280
 :  60505-01
Decon 90 Cleaning Agent, Electron Microscopy Sciences
  102093-280
Decon 90 Cleaning Agent, Electron Microscopy Sciences
Decon 90 Cleaning Agent, Electron Microscopy Sciences

Decon 90, 1 l, Bottle

 
$87.42
   
Each
 
   
 

Decon 90 is the leading surface active cleaning agent, and/or radioactive decontaminant, for laboratory, medical and specialized industrial applications. It can be used for ʽmanual’ cleaning or in an ultrasonic cleaning bath.

  • Biodegradable *, phosphate-free, Bactericidal **, non-flammable and totally rinsable
  • NOT suitable for domestic applications
  • Supplied as a liquid concentrate, for dilution with water

Appearance: Milky white, non-viscous liquid, having faint odour. pH: DECON 90 is alkaline. The concentrate having a pH in excess of pH 13.

Applications: For cleaning and/or decontaminating glassware, ceramics, rubbers, plastics, stainless steel and ferrous metals. Being alkaline, DECON 90 is NOT suitable for use on non-ferrous metals, notably aluminium and zinc, or on polycarbonate.

Formulation: An emulsion of anionic and non-ionic surface active agents, stabilizing agents, non-phosphate detergent builders, alkalis and sequestering agents, in an aqueous base. Decon 90 does NOT contain Phosphates, Enzymes, EDTA/NTA or Chlorine Bleaches.

 : Certificates of analysis are available.

 : Supplied in high density polyethylene containers, with tamper-evident, screw-on caps.

 : ** Decon 90 satisfies the criteria in BS EN 1040 1997 for bacterial activity. * Ready Biodegradability by OECD 301E (ISO method 7827 - 1986 (E).

 
  • Description:
    Decon 90
  • Size:
    1 l
  • Packaging:
    Bottle

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.