MicroPlate Genie®, Electron Microscopy Sciences
High Speed Action For Increased Mixing Efficiency In Every Well!
- Aggressive mixing speed from 750-3200 rpm for complete and uniform mixing in any microplate format
- May be used to shake any single microplate
- Multi capacity microplate mixer is also available (Multi-MicroPlate Genie)
- Small vortexing orbit of 1.0 mm for thorough mixing regardless of sample viscosity
- Speed and orbit combine to offer true vortexing action in each well of a microplate
- Accepts most microplate types within the recommendations of the Society for Biomolecular Screening (SBS), even in 384-well formats
- Built-in timer operates from 1 to 60 minutes or continuous for hands-free use
- May be used in cold rooms and incubators for temperature sensitive applications
Additional accessories for the Microplate Genie® available.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.