Orders LinkContactUsLinkComponent
Waterproof Labels, Wet Grip, Electron Microscopy Sciences
Waterproof Labels, Wet Grip, Electron Microscopy Sciences
Certificates
Waterproof Labels, Wet Grip, Electron Microscopy Sciences
Waterproof Labels, Wet Grip, Electron Microscopy Sciences
10 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Labels for moist, frosty or frozen tubes.
  • Temperature resistant to –40 °C
Direct thermal Labels are white and on a 1" int.Ø. core. Temperature Range: –40 to +70 °C.
GRIPS to wet, frost-covered and frozen surface, Compatible with centrifuge rotors, Resistant to chemicals and solvents, Adheres to plastics, glass, metals and even paperboard boxes.
Print on sheets with Laser printers or use rolls for direct thermal printers.
Product Family Options
Product Information
  • For Use With
    Diameter
    Dimensions
    Availability
    Price
  • 102098-712
    Waterproof labels, wet grip
    Waterproof labels, wet grip
    1.5 ml tubes
    9.5 mm (0.37")
    Log in for availability
    Log in for availability
    Roll of 1,000
    Roll of 1,000 - $119.53
    $119.53
    Roll of 1,000
       $119.53
    Supplier Number: 77594-60
  • 102681-382
    Waterproof labels, wet grip
    Waterproof labels, wet grip
    0.5 ml tubes
    9.5 mm (0.37")
    Log in for availability
    Log in for availability
    Pack of 1,000
    Pack of 1,000 - $133.98
    $133.98
    Pack of 1,000
       $133.98
    Supplier Number: 77594-40
  • 102681-390
    Waterproof labels, wet grip
    Waterproof labels, wet grip
    Microscope slides
    22 x 22 mm
    Log in for availability
    Log in for availability
    Pack of 1,000
    Pack of 1,000 - $143.99
    $143.99
    Pack of 1,000
       $143.99
    Supplier Number: 77595-40
  • 102681-386
    Waterproof labels, wet grip
    Waterproof labels, wet grip
    0.5 ml tubes
    24 x 13 mm
    Log in for availability
    Log in for availability
    Pack of 2,380
    Pack of 2,380 - $143.99
    $143.99
    Pack of 2,380
       $143.99
    Supplier Number: 77595-20
  • 102098-722
    Waterproof labels, wet grip
    Waterproof labels, wet grip
    1.5 ml tubes
    33 x 13 mm
    Log in for availability
    Log in for availability
    Roll of 1,000
    Roll of 1,000 - $71.97
    $71.97
    Roll of 1,000
       $71.97
    Supplier Number: 77595-60
  • 102098-724
    Waterproof labels, wet grip
    Waterproof labels, wet grip
    1.5 - 2 ml tubes
    33 x 13 mm
    Log in for availability
    Log in for availability
    Roll of 1,000
    Roll of 1,000 - $54.28
    $54.28
    Roll of 1,000
       $54.28
    Supplier Number: 77595-65
  • 102681-384
    Waterproof labels, wet grip
    Waterproof labels, wet grip
    1.5 ml tubes
    33 x 13 mm
    Log in for availability
    Log in for availability
    Pack of 1,700
    Pack of 1,700 - $143.99
    $143.99
    Pack of 1,700
       $143.99
    Supplier Number: 77595-10
  • 102098-704
    Waterproof labels, wet grip
    Waterproof labels, wet grip
    1.5 ml tubes
    38 x 13 mm
    Log in for availability
    Log in for availability
    Roll of 1,000
    Roll of 1,000 - $102.81
    $102.81
    Roll of 1,000
       $102.81
    Supplier Number: 77594-20
  • 102098-706
    Waterproof labels, wet grip
    Waterproof labels, wet grip
    2 ml tubes
    38 x 19 mm
    Log in for availability
    Log in for availability
    Roll of 1,000
    Roll of 1,000 - $88.59
    $88.59
    Roll of 1,000
       $88.59
    Supplier Number: 77594-30
  • 102681-388
    Waterproof labels, wet grip
    Waterproof labels, wet grip
    1.5 - 2 ml tubes
    38 x 19 mm
    Log in for availability
    Log in for availability
    Pack of 1,000
    Pack of 1,000 - $143.99
    $143.99
    Pack of 1,000
       $143.99
    Supplier Number: 77595-30
  • Discontinued
    Supplier Number: -
  • Discontinued
    Supplier Number: -