About this item
A Heyman type micro spatula made from nickel stainless steel with a glazed finish.
- Wire Ø - 2.62 mm (0.103")
- Overall length: 159 mm (6¹/₄")
- Flat end: 31.35 mm (1¹/₄") long
The spoon end: 12.5 mm (½") long × 3 mm (¹/₈") wide. V-Shaped formed other end: ½" long.