Orders LinkContactUsLinkComponent
Strip Disposable Mold, Electron Microscopy Sciences
Strip Disposable Mold, Electron Microscopy Sciences
Certificates
Strip Disposable Mold, Electron Microscopy Sciences
Strip Disposable Mold, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Strip disposable molds are manufactured from high impact polystyrene, offering a low cost alternative to stainless steel
These molds are available in a choice of sizes and depths. The molds have a smooth interior finish and rounded corners for ease of removal. There are five molds per strip and they are packed 100 strips to a case.
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 30 x 24 x 5 mm
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $115.77
    $115.77
    Pack of 100
       $115.77
    Supplier Number: 62351-30
  • 37 x 24 x 7 mm
    Discontinued
    Supplier Number: -